Learn More
DHRS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20172-1-AP-150UL
 View more versions of this product
Product Code. 16889674
  • 4652.90 SEK / 150µL
Description
DHRS11 gene ontology annotations related to this gene include oxidoreductase activity.
Specifications
Specifications
DHRS11
Polyclonal
Unconjugated
DHRS11
ARPG836, DHRS11, SDR24C1
Rabbit
Antigen Affinity Chromatography
RUO
192970, 360583, 79154
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3U0B3, Q6UWP2
Dhrs11
DHRS11 Fusion Protein Ag14005
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DHRS11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated