missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHRS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DHRS2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895044
|
Proteintech
15735-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885044
|
Proteintech
15735-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.Specifications
DHRS2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q13268 | |
10202 | |
DHRS2 Fusion Protein Ag8508 | |
Primary | |
-20°C | |
DHRS2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DHRS2, Dicarbonyl reductase HEP27, HEP27, Protein D, SDR25C1 | |
DHRS2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DHRS2 Rabbit anti-Human, Polyclonal, Proteintech