Learn More
DHRS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15279-1-AP-150UL
 View more versions of this product
Product Code. 16884384
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Reduces all-trans-retinal and 9-cis retinal. Can also catalyze the oxidation of all-trans-retinol with NADP as co-factor, but with much lower efficiency. Reduces alkyl phenyl ketones and alpha-dicarbonyl compounds with aromatic rings, such as pyrimidine-4-aldehyde, 3-benzoylpyridine, 4-benzoylpyridine, menadione and 4-hexanoylpyridine. Has no activity towards aliphatic aldehydes and ketones.
Specifications
Specifications
DHRS4
Polyclonal
Unconjugated
DHRS4
CR, DHRS4, DHRS4L2, humNRDR, NRDR, PHCR, PSCD, SCAD SRL, SDR SRL, SDR25C1
Rabbit
Antigen Affinity Chromatography
RUO
10901, 266686, 28200
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8VID1, Q99LB2, Q9BTZ2
Dhrs4
DHRS4 Fusion Protein Ag7398
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DHRS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated