Learn More
DHX15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12265-1-AP-20UL
 View more versions of this product
Product Code. 16879873
  • 1645.54 SEK / 20µL
Description
HMGB3 (HMG4) belongs to the high mobility group protein superfamily. Like HMG1 and HMG2 , HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.
Specifications
Specifications
DHX15
Polyclonal
Unconjugated
DHX15
ATP dependent RNA helicase 46, DBP1, DDX15, DEAH box protein 15, DHX15, HRH2, PRP43, PRPF43, PrPp43p
Rabbit
Antigen Affinity Chromatography
RUO
13204, 1665, 289693
-20°C
Liquid
Western Blot, Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35286, O43143
Dhx15
DHX15 Fusion Protein Ag2895
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DHX15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated