Läs mer
Diacerein, Thermo Scientific™
Interleukin-1 inhibitor
2069.79 SEK - 5541.71 SEK
Kemiska identifierare
| CAS | 13739-02-1 |
|---|---|
| Molekylformel | C19H12O8 |
| Molekylvikt (g/mol) | 368.30 |
| MDL-nummer | MFCD00468030 |
| InChI-nyckel | TYNLGDBUJLVSMA-UHFFFAOYSA-N |
| Synonym | diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french |
| PubChem CID | 26248 |
| IUPAC-namn | 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid |
| LEDER | CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
15415949
|
Thermo Scientific Alfa Aesar
J63775.03 |
1g |
2069.79 SEK
1g |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
15425949
|
Thermo Scientific Alfa Aesar
J63775.06 |
5g |
5541.71 SEK
5g |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiska identifierare
| 13739-02-1 | |
| 368.30 | |
| TYNLGDBUJLVSMA-UHFFFAOYSA-N | |
| 26248 | |
| CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O |
| C19H12O8 | |
| MFCD00468030 | |
| diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french | |
| 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid |
Specifikationer
| Diacerein | |
| C19H12O8 | |
| 14,2960 | |
| TYNLGDBUJLVSMA-UHFFFAOYSA-N | |
| 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid | |
| 26248 |
| 13739-02-1 | |
| MFCD00468030 | |
| diacerein, diacetylrhein, diacerhein, artrodar, fisiodar, 1,8-diacetoxy-3-carboxyanthraquinone, 4,5-diacetylrhein, diacerin, diacerein inn, diacereine french | |
| CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O | |
| 368.30 | |
| 368.29 |
Säkerhet och hantering
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 237-310-2
missing translation for 'rtecsNumber' : CB8781000
missing translation for 'tsca' : No
Rekommenderad förvaring : Ambient temperatures