Läs mer
Diacetyl 2,4-Dinitrophenylhydrazone, TRC
Produktkod. 30007202
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007202 1 g
30003304 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30007202 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4225501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2256-00-0
Molekylformel C10H10N4O5
Molekylvikt (g/mol) 266.21
Synonym 2-[2-(2,4-Dinitrophenyl)hydrazone] 2,3-Butanedione, Mono[(2,4-dinitrophenyl)
IUPAC-namn 3-(2-(2,4-dinitrophenyl)hydrazineylidene)butan-2-one
LEDER O=C(C)/C(C)=N/NC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1
Specifikationer
Kemiskt namn eller material Diacetyl 2,4-Dinitrophenylhydrazone
Kvantitet 1 g
InChI formel InChI=1S/C10H10N4O5/c1-6(7(2)15)11-12-9-4-3-8(13(16)17)5-10(9)14(18)19/h3-5,12H,1-2H3/b11-6+
Formel vikt 266.07