Learn More
Thermo Scientific™ Viper™ Automated Method Scouting Solution Kits
Obtain all the easy, zero-dead-volume fittings and capillaries needed for your method scouting LC system in this single kit.
Description
Avoid data-destroying dead volumes in the capillary connections of your method scouting HPLC or UHPLC system by using the Thermo Scientific™ Viper™ Method Scouting Solutions kit. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system. A single kit contains all the connectors and capillaries you need and ensures consistently perfect connections.
Specifications
Automated Method Scouting Solution Kit |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.