missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIRAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DIRAS1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850454
|
Proteintech
12634-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840454
|
Proteintech
12634-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.Specifications
DIRAS1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O95057, Q91Z61 | |
148252, 208666, 366826 | |
DIRAS1 Fusion Protein Ag3329 | |
Primary | |
-20°C | |
DIRAS1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Di Ras1, DIRAS1, GBTS1, GTP binding protein Di Ras1, RIG | |
Diras1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DIRAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech