missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21599-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to DNA2.
DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.Specifications
DNA2 | |
Polyclonal | |
Unconjugated | |
DNA2 | |
DNA2, DNA2L, KIAA0083 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1763 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51530 | |
DNA2 | |
DNA2 Fusion Protein Ag16206 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNA2 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction