missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DNAH6 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898114
|
Proteintech
18080-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888114
|
Proteintech
18080-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to DNAH6.
DNAH6 is a protein coding gene.Specifications
DNAH6 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9C0G6 | |
117250, 1768, 330355 | |
DNAH6 Fusion Protein Ag12774 | |
Primary | |
-20°C | |
DNAH6 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Ciliary dynein heavy chain 6, DNAH6, Dnahc6, DNHL1, Dynein heavy chain 6, axonemal, HL 2, HL2, KIAA1697 | |
Dnah6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNAH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech