missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC2/MPP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11971-1-AP-150UL
This item is not returnable.
View return policy
Description
EIF4EBP1 is one member of a family of translation repressor proteins. It directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5′ end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.Specifications
DNAJC2/MPP11 | |
Polyclonal | |
Unconjugated | |
DNAJC2 | |
DNAJC2, M phase phosphoprotein 11, MPP11, ZRF1, ZUO1, Zuotin related factor 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116456, 22791, 27000 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54103, Q7TQ20, Q99543 | |
DNAJC2 | |
DNAJC2/MPP11 Fusion Protein Ag2594 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNAJC2/MPP11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction