missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DNAJC21 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882335
|
Proteintech
23411-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872335
|
Proteintech
23411-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May act as a co-chaperone for HSP70. May play a role in ribosomal RNA (rRNA) biogenesis, possibly in the maturation of the 60S subunit. Binds the precursor 45S rRNA.Specifications
DNAJC21 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
E9Q8D0, Q5F1R6 | |
134218, 192210, 78244 | |
DNAJC21 Fusion Protein Ag20003 | |
Primary | |
-20°C | |
DNAJC21 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DNAJA5, DNAJC21, GS3, JJJ1, Protein GS3 | |
Dnajc21 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNAJC21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech