missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNASE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DNASE2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895334
|
Proteintech
15934-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885334
|
Proteintech
15934-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DNase II is a member of the DNase family. The protein, located in the lysosome, hydrolyzes DNA under acidic conditions and mediates the breakdown of DNA during erythropoiesis and apoptosis. Two codomit alleles have been characterized, DNASE2*L (low activity) and DNASE2*H (high activity), that differ at one nucleotide in the promoter region. The DNASE2*H allele is represented in this record.Specifications
DNASE2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O00115, P56542, Q9QZK8 | |
13423, 171575, 1777 | |
DNASE2 Fusion Protein Ag8781 | |
Primary | |
-20°C | |
DNASE2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Acid DNase, Deoxyribonuclease 2 alpha, Deoxyribonuclease II alpha, DNase II alpha, DNASE2, DNASE2A, DNL, DNL2, Lysosomal DNase II, R31240_2 | |
Dnase2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DNASE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech