Learn More
DOCK7 Mouse, anti-Human, Unconjugated, Clone: 1A4A6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67842-1-IG-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17221506
  • 4721.67 SEK / 150µL
Estimated Shipment: 20-05-2024
to see stock.
Description
Description
DOCK 7 (dedicator of cytokinesis 7), also known as ZIR2, is a member of the DOCK180-related protein superfamily. Expressed mainly in neuronal cells, DOCK 7 is a guanine nucleotide exchange factor (GEF) for small GTPases, Rac1 and Cdc42, which are the major regulators of actin cytoskeleton. Multiple isoforms of DOCK 7 exist due to alternative splicing events.
Specifications
Specifications
DOCK7
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, ELISA
1A4A6
Unconjugated
150 μL
85440