Learn More
DOCK7 Mouse, anti-Human, Unconjugated, Clone: 1A4A6, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DOCK7 |
---|---|
Clone | 1A4A6 |
Concentration | 500 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17231506
|
Proteintech
67842-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17221506
|
Proteintech
67842-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DOCK 7 (dedicator of cytokinesis 7), also known as ZIR2, is a member of the DOCK180-related protein superfamily. Expressed mainly in neuronal cells, DOCK 7 is a guanine nucleotide exchange factor (GEF) for small GTPases, Rac1 and Cdc42, which are the major regulators of actin cytoskeleton. Multiple isoforms of DOCK 7 exist due to alternative splicing events.
Specifications
DOCK7 | |
500 ÎĽg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat | |
Primary |
1A4A6 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
85440 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.