Läs mer
Doxycycline monohydrate, Thermo Scientific™
Doxycycline monohydrate, CAS # 17086-28-1, is the hydrated form of the tetracycline class antibiotic doxycycline.
Brand:  Thermo Scientific Alfa Aesar J63805.06
871.23 SEK valid until 2026-09-25
BÄSTA PRIS på kampanj! Använd kampanjkod: "27966" för att få ditt kampanjpris.
Kvantitet 5g
 Se fler versioner av denna produkt
Produktkod. 15435979
  • ONLINE EXCLUSIVE PRICE
    1024.88 SEK
    919.65 SEK / 5g
    Save 105.23 SEK
    10% Av
Visa alla kampanjer
 Begränsad artikel
I lager
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
• It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
• Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
• Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties
Application
• Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
• Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vivo and in vitro
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiska identifierare
17086-28-1
462.455
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
C22H26N2O9
MFCD02682958
54684461
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
Specifikationer
Doxycycline monohydrate
Yellow
C22H26N2O9
14,3440
FZKWRPSUNUOXKJ-CVHRZJFOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
54684461
17086-28-1
5g
MFCD02682958
Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide.
CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O
462.455
462.46 (444.44 Anhydrous)
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Promo
Utgår: 2026-09-25
Nyfikenheten går längre
Kampanjkod: 27966
Flytta gränserna för forskning med besparingar inom forskningsarbetsflöden: Cellodling och analys med vår Gibco™-produktportfölj, genuttryck med Applied Biosystems™ samt proteinuttryck med Invitrogen™ och Thermo Scientific™.
Tillämpa kampanj och lägga till...