missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12222-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a kidney membrane enzyme involved in the metabolism of glutathione and other similar proteins by dipeptide hydrolysis. The encoded protein is known to regulate leukotriene activity by catalyzing the conversion of leukotriene D4 to leukotriene E4. This protein uses zinc as a cofactor and acts as a disulfide-linked homodimer. Two transcript variants encoding the same protein have been found for this gene.Specifications
DPEP1 | |
Polyclonal | |
Unconjugated | |
DPEP1 | |
Dehydropeptidase I, Dipeptidase 1, dipeptidase 1 (renal), DPEP1, hRDP, MBD1, MDP, Microsomal dipeptidase, RDP, Renal dipeptidase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13479, 1800, 94199 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16444, P31428, P31430 | |
DPEP1 | |
DPEP1 Fusion Protein Ag2907 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DPEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction