missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12111-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the d4 domain family, characterized by a zinc finger-like structural motif. This protein functions as a transcription factor which is necessary for the apoptotic response following deprivation of survival factors. It likely serves a regulatory role in rapid hematopoietic cell growth and turnover. This gene is considered a candidate gene for multiple endocrine neoplasia type I, an inherited cancer syndrome involving multiple parathyroid, enteropancreatic, and pituitary tumors.Specifications
DPF2 | |
Polyclonal | |
Unconjugated | |
DPF2 | |
BAF45D, BRG1 associated factor 45D, DPF2, Protein requiem, REQ, ubi d4, UBID4, Zinc finger protein ubi d4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19708, 361711, 5977 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q61103, Q92785 | |
Dpf2 | |
DPF2 Fusion Protein Ag2756 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |