Learn More
DRD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17934-1-AP-150UL
 View more versions of this product
Product Code. 16887944
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
Specifications
Specifications
DRD1
Polyclonal
Unconjugated
DRD1
D(1A) dopamine receptor, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A
Rabbit
Antigen Affinity Chromatography
RUO
13488, 1812
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21728, Q61616
DRD1
DRD1 Fusion Protein Ag12366
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DRD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated