missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DRD5 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879834
|
Proteintech
20310-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869834
|
Proteintech
20310-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.Specifications
DRD5 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P21918, P25115, Q8BLD9 | |
13492, 1816, 25195 | |
DRD5 Fusion Protein Ag14145 | |
Primary | |
-20°C | |
DRD5 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D(1B) dopamine receptor, D(5) dopamine receptor, D1beta dopamine receptor, DBDR, Dopamine D5 receptor, dopamine receptor D5, DRD1B, DRD1L2, DRD5 | |
Drd5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DRD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech