Learn More
Applied Biosystems™ DS-32 Matrix Standard Set [5FAM™, JOE™, NED™ and ROX™ dyes] for the 310 Genetic Analyzer
Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples.
Brand: Applied Biosystems™ 4312131
Includes: Contains dye-labeled oligonucleotides used to generate the multicomponent matrix required for the AmpF/STR® kit.
Description
The matrix standard st is used to generate the 'multicomponent matrix' required when analyzing DNA fragments on the 310 genetic anlayzer.Order Info
Kit
Specifications
Contains dye-labeled oligonucleotides used to generate the multicomponent matrix required for the AmpF⁄STR™ kit. Store at 2 - 8°C. Do not freeze. | |
Sequencing | |
5FAM™, JOE™, NED™, ROX™ | |
1 Kit |
310 Genetic Analyzer | |
FAM, JOE, NED, ROX | |
DS-32 Matrix Standard Set | |
Wet Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2006165 | Certificate of Analysis | 22/06/2020 | 4312131 |
1001132 | Certificate of Analysis | 07/01/2019 | 4312131 |
1805163 | Certificate of Analysis | 30/05/2018 | 4312131 |
1404156 | Certificate of Analysis | 19/07/2017 | 4312131 |
1703162 | Certificate of Analysis | 19/07/2017 | 4312131 |
For research use only