Learn More
Macherey-Nagel™ Duotest™ pH Paper Roll
Product Code. 10291991 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
1 to 4.3
3.5 to 6.8
5 to 8
7 to 10
9.5 to 14
Unit Size:
5m
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10291991 5 to 8 Each
10084880 1 to 4.3 5m
10320771 3.5 to 6.8 5m
10360151 7 to 10 5m
10094880 9.5 to 14 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10291991 Supplier Macherey-Nagel™ Supplier No. 90304
  • ONLINE EXCLUSIVE PRICE
    257.00 SEK
    181.39 SEK / Paket med 1
    Save 75.61 SEK
    29% Av
 Restricted Item
I lager
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
  • Provides rapid and reliable results
  • Available for a range of pH gradations
  • Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
  • Allows more accurate reading and good estimation of intermediate values
Specifications
pH Range 5 to 8
Format Roll
Graduations 5,0, 5,3, 5,6, 5,9, 6,2, 6,5, 6,8, 7,1, 7,4, 7,7, 8,0
Quantity 1 Each
Type pH Paper
Length (Metric) 5 m
Width (Metric) 10 mm