Learn More
DWK Life Sciences DURAN™ Rolled Flange Bottle, without closure
Alle ansehen DWK Life Sciences Produkte
missing translation for 'orderingAttributeHoverText'
Kapacitet (metrisk):
100 mL
250 mL
500 mL
1000 mL
missing translation for 'unitSize'
Paket med 10
Beschreibung
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. While some bottles and flasks are used for the cultivation of micro-organisms, the roller rim bottles are mainly used for sampling (e.g. in the dairy industry).
- Supplied without screw cap
Spezifikation
Spezifikation
| Kapacitet (metrisk) | 250 mL |
| För användning med (applikation) | Sample collection; Storage and transport of Liquid cultures; Media preparation and cultivation; Delivery without closure |
| Material | Borosilicate Glass (3.3) |
| Kvantitet | 10/Pk. |
| Diameter (metrisk) | 57 mm |
| Autoklaveringsbar | Autoclavable |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur