missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | DUSP13 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887603
|
Proteintech
10909-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877603
|
Proteintech
10909-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dual-specificity phosphatases, a subfamily of protein-tyrosine phosphatases, play important roles in signal transduction, cell cycle progression, and tumor suppression. The cDNA encoding a novel phosphatase, PIR1, phosphatase that interacts with RNA/RNP complex 1. Sequence analysis revealed that the predicted 329-amino acid protein has homology to several dual-specificity phosphatases and contains 2 stretches of arginine-rich sequence similar to those found in some RNA-binding proteins. In vitro, recombit protein displays protein-tyrosine phosphatase activity and binds directly to RNA.Specifications
DUSP13 | |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q6B8I0, Q6B8I1 | |
27389, 361002, 51207 | |
DUSP13 Fusion Protein Ag1348 | |
Primary | |
-20°C | |
DUSP13 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BEDP, DUSP13, DUSP13A, DUSP13B, MDSP, SKRP4, TMDP | |
Dusp13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DUSP13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech