Learn More
3M™ Dustmaster™ Particulate Filters Product Code.: 15729797
3M™ Dustmaster™ Particulate Filters
Product Code. 15729797
P3, 5/Pk., Each
Click to view available options
Filter Type:
P2
P2R
P3
Quantity:
1 Ea.
5/Pk.
Unit Size:
10 pairs
Each
Product Code. 15729797
Brand: 3M™ 7000059866
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
  • Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
  • Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
TRUSTED_SUSTAINABILITY
Specifications
Filter Type P3
Product Type Dust Filter
For Use With (Equipment) 6000 and 7000 series facemasks
Includes Filter
Quantity 5/Pk.
Product Content Correction
Product Title
3M™ Dustmaster™ Particulate Filters >