Learn More
3M™ Dustmaster™ Particulate Filters
Click to view available options
Filter Type:
P2
P2R
P3
Quantity:
1 Ea.
5/Pk.
Unit Size:
10 pairs
Each
Description
- Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
- Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
Specifications
Filter Type | P2 |
Product Type | Dust Filter |
For Use With (Equipment) | 6000 and 7000 series facemasks |
Includes | Gasket |
Quantity | 1 Ea. |
Product Content Correction
Product Title
3M™ Dustmaster™ Particulate Filters >
Spot an opportunity for improvement?Share a Content Correction