Learn More
ABEBA™ Dynamic Flow Black Occupational Slip-On O1
Occupational slip-on shoes, breathable black knitted fabric, replaceable active MediFit insole, slip resistant TPU outsole
Brand: ABEBA™ 7376/46
This item is not returnable.
View return policy
Description
- Breathable, lightweight, comfortable
- Comply to EN ISO 20347: 2012, O1 SRC
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Black knitted fabric breathable
- Double elastic band fitting system
- Removable active Medifit insole
Specifications
Black | |
46 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
Comply to EN ISO 20347: 2012 O1 SRC | |
Slip-on | |
O1 SRC |
Product Content Correction
Product Title
ABEBA™ Dynamic Flow Black Occupational Slip-On O1 > Size: 46
Spot an opportunity for improvement?Share a Content Correction