missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC1LI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18885-1-AP-20UL
This item is not returnable.
View return policy
Description
Cytoplasmic dynein is a microtubule-associated motorprotein.Specifications
DYNC1LI2 | |
Polyclonal | |
Unconjugated | |
DYNC1LI2 | |
DNCLI2, DYNC1LI2, LIC 2, LIC2, LIC53/55 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1783, 234663, 81655 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43237, Q62698, Q6PDL0 | |
DYNC1LI2 | |
DYNC1LI2 Fusion Protein Ag13452 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DYNC1LI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction