missing translation for 'onlineSavingsMsg'
Learn More
Invitrogen™ Human DYRK1A, GST Tag Recombinant Protein Product Code.: 11563527

Invitrogen™ Human DYRK1A, GST Tag Recombinant Protein

Product Code. p-7075834
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 11563527

Brand: Invitrogen™ PV3785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease. DYRK1A catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13627
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1859
Molecular Weight (g/mol) 113 kDa
Name Human DYRK1A, GST Tag
Quantity 10 μg
Gene Alias 2310043O08Rik; D16Ertd272e; D16Ertd493e; dual specificity tyrosine phosphorylation regulated kinase 1 A; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1 A; dual specificity tyrosine-phosphorylation-regulated kinase 1 A; Dual specificity YAK1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1 A; Dyrk; DYRK1; Dyrk1a; ENSMUSG00000074897; Gm10783; hMNB; HP86; mmb; MNB; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; MRD7; OTTHUMP00000109090; Protein kinase minibrain homolog; PSK47; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Common Name DYRK1A
KinaseFamily DYRK Kinase Family, DYRKs
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CMCG Group
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™
Target Entry DYRK1A
Target Gene dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1 A
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Genetic Diseases
Purity or Quality Grade 85% by SDS-PAGE
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number2639089D Certificate TypeCertificate of Analysis Date01/11/2023 Product CodePV3785
Lot Number2639089B Certificate TypeCertificate of Analysis Date28/04/2023 Product CodePV3785
Lot Number2502867B Certificate TypeCertificate of Analysis Date02/11/2022 Product CodePV3785
Lot Number1783084P Certificate TypeCertificate of Analysis Date18/04/2022 Product CodePV3785
Lot Number2427476C Certificate TypeCertificate of Analysis Date06/04/2022 Product CodePV3785
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human DYRK1A, GST Tag Recombinant Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.