Learn More
Invitrogen™ eBioscience™ Avidin HRP
Commonly used as a second step for the detection of biotinylated antibodies. Avidin is a glycoprotein from chicken egg whites that binds biotin with high affinity. Biotin is a tag that is frequently used to label antibodies and other probes for immunodetection methods.
830.00 SEK - 2490.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15558306
|
Invitrogen™
18-4200-93 |
100μL |
830.00 SEK
100µL |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15548306
|
Invitrogen™
18-4200-89 |
5 x 100μL |
2490.00 SEK
Pack of 5 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Recommended for use in immunohistochemistry applications
- Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be used with this product
- 0.5mg/mL
For Research Use Only. Not for use in diagnostic procedures.