Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 45 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15484
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12498230
  • 1425.00 SEK / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket
Description
Description
  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
5 x 5
Aluminum
450 x 135 x 233 mm
Microtiterplates (25)
Upright Freezers
45 mm