missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 45 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
Marque: Tenak™ TE15540
Informations supplémentaires : Weight : 1.60000kg
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Spécification
3 x 6 | |
Aluminum | |
540 x 135 x 140 mm |
Microtiterplates (18) | |
Upright Freezers | |
45 mm |