missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECOP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12611-1-AP-20UL
This item is not returnable.
View return policy
Description
Increases the transcriptional activity of NFKB1 by facilitating its nuclear translocation, DNA-binding and associated apoptotic response, when overexpressed.Specifications
ECOP | |
Polyclonal | |
Unconjugated | |
VOPP1 | |
ECOP, GASP, VOPP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
232023, 362374, 81552 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B5DEK8, Q8R1C3, Q96AW1 | |
Vopp1 | |
ECOP Fusion Protein Ag3225 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ECOP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction