Learn More
EFHD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12181-1-AP-150UL
 View more versions of this product
Product Code. 16889733
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
EFHD2, also known as Swiprosin-1 or SWS1, is an EF-hand and coiled-coil-containing adaptor protein that plays a role in lymphocyte physiology. EFHD2 exhibits the highest expression in CD8+ T cells and immature B cells. It provides a membrane scaffold that is required for the Syk-, SLP-65-, and PLC-gamma2-dependent B-cell receptor (BCR)-induced calcium flux. EFHD2 may also regulate BCR-induced immature and primary B-cell apoptosis. It controls spontaneous apoptosis through the regulation of BCL2L1 abundance. Also, EFHD2 plays a role as negative regulator of the canonical NF-kappa-B-activating branch.
Specifications
Specifications
EFHD2
Polyclonal
Unconjugated
EFHD2
EFHD2, Swiprosin 1, SWS1
Rabbit
Antigen Affinity Chromatography
RUO
27984, 79180
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96C19, Q9D8Y0
Efhd2
EFHD2 Fusion Protein Ag2826
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EFHD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated