missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22008-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.Specifications
EGR1 | |
Polyclonal | |
Unconjugated | |
EGR1 | |
AT225, early growth response 1, EGR 1, EGR1, G0S30, KROX 24, KROX24, NGFI A, TIS8, Transcription factor ETR103, Transcription factor Zif268, ZIF 268, Zinc finger protein 225, Zinc finger protein Krox 24, ZNF225 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13653, 1958 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08046, P18146 | |
EGR1 | |
EGR1 Fusion Protein Ag13395 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EGR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction