missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
EGR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22008-1-AP-150UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.Specifikationer
| EGR1 | |
| Polyclonal | |
| Unconjugated | |
| EGR1 | |
| AT225, early growth response 1, EGR 1, EGR1, G0S30, KROX 24, KROX24, NGFI A, TIS8, Transcription factor ETR103, Transcription factor Zif268, ZIF 268, Zinc finger protein 225, Zinc finger protein Krox 24, ZNF225 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13653, 1958 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08046, P18146 | |
| EGR1 | |
| EGR1 Fusion Protein Ag13395 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion