missing translation for 'onlineSavingsMsg'
Learn More

EHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11440-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828523

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the EH domain-containing protein family. These proteins are characterized by a C-terminal EF-hand domain, a nucleotide-binding consensus site at the N terminus and a bipartite nuclear localization signal. The encoded protein interacts with the actin cytoskeleton through an N-terminal domain and also binds to an EH domain-binding protein through the C-terminal EH domain. This interaction appears to connect clathrin-dependent endocytosis to actin, suggesting that this gene product participates in the endocytic pathway.
Specifications

Specifications

EHD2
Polyclonal
Unconjugated
EHD2
EH domain containing 2, EH domain containing protein 2, EHD2, PAST homolog 2, PAST2
Rabbit
Antigen Affinity Chromatography
RUO
259300, 30846, 361512
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4V8H8, Q8BH64, Q9NZN4
EHD2
EHD2 Fusion Protein Ag2002
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers