missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | EHD2 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828523
|
Proteintech
11440-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818523
|
Proteintech
11440-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the EH domain-containing protein family. These proteins are characterized by a C-terminal EF-hand domain, a nucleotide-binding consensus site at the N terminus and a bipartite nuclear localization signal. The encoded protein interacts with the actin cytoskeleton through an N-terminal domain and also binds to an EH domain-binding protein through the C-terminal EH domain. This interaction appears to connect clathrin-dependent endocytosis to actin, suggesting that this gene product participates in the endocytic pathway.Specifications
EHD2 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q4V8H8, Q8BH64, Q9NZN4 | |
259300, 30846, 361512 | |
EHD2 Fusion Protein Ag2002 | |
Primary | |
-20°C | |
EHD2 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
EH domain containing 2, EH domain containing protein 2, EHD2, PAST homolog 2, PAST2 | |
EHD2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |