Learn More
EIF1 Mouse anti-Human, Unconjugated, Clone: 2G8C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67601-1-IG-150UL
This item is not returnable.
View return policy
Description
The EIF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF1. This antibody recognizes human antigen. The EIF1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
EIF1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
2G8C6 | |
Unconjugated | |
150 μL | |
10209 | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EIF1 Mouse anti-Human, Unconjugated, Clone: 2G8C6, Proteintech > 150 μL
Spot an opportunity for improvement?Share a Content Correction