Learn More
EIF1 Mouse anti-Human, Unconjugated, Clone: 2G8C6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67601-1-IG-150UL
 View more versions of this product
Product Code. 16890316
  • 4652.90 SEK / 150µL
Description
The EIF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF1. This antibody recognizes human antigen. The EIF1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
EIF1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
2G8C6
Unconjugated
150 μL
10209
IgG2b
Certificates
Promotions
Promotions
Product Content Correction
Product Title
EIF1 Mouse anti-Human, Unconjugated, Clone: 2G8C6, Proteintech > 150 μL