missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10463-1-AP-20UL
This item is not returnable.
View return policy
Description
eIF4A3 is a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. eIF4A3 is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family.Specifications
EIF4A3 | |
Polyclonal | |
Unconjugated | |
EIF4A3 | |
DDX48, DEAD box protein 48, eIF 4A III, eIF4A III, EIF4A3, eIF4AIII, hNMP 265, hNMP265, KIAA0111, Nuclear matrix protein 265, NUK 34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192170, 9775 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P38919, Q91VC3 | |
EIF4A3 | |
EIF4A3 Fusion Protein Ag0733 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EIF4A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction