missing translation for 'onlineSavingsMsg'
Learn More

ELF1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1662.46 SEK - 4700.76 SEK

Specifications

Antigen ELF1
Concentration 0.46 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896465
missing translation for 'viewDocuments'
Proteintech
55029-1-AP-20UL
20 ÎĽL
1662.46 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16886465
missing translation for 'viewDocuments'
Proteintech
55029-1-AP-150UL
150 ÎĽL
4700.76 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody is specific to ELF1.

This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ELF1
Western Blot
Unconjugated
Rabbit
Human
P32519
1997
Peptide
Primary
-20°C
ELF1
0.46 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
E74 like factor 1, ELF1
ELF1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ELF1 Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.