missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELMOD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12761-1-AP-20UL
This item is not returnable.
View return policy
Description
Acts as a GTPase-activating protein (GAP) toward guanine nucleotide exchange factors like ARL2, ARL3, ARF1 and ARF6, but not for GTPases outside the Arf family.Specifications
ELMOD1 | |
Polyclonal | |
Unconjugated | |
ELMOD1 | |
ELMOD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55531 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N336 | |
ELMOD1 | |
ELMOD1 Fusion Protein Ag3368 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELMOD1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction