missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELMOD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13027-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of six engulfment and motility (ELMO) domain-containing proteins. This gene is thought to play a role in antiviral responses. Mutations in this gene may be involved in the cause of familial idiopathic pulmonary fibrosis.Specifications
ELMOD2 | |
Polyclonal | |
Unconjugated | |
ELMOD2 | |
9830169G11Rik, ELMOD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
255520 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IZ81 | |
ELMOD2 | |
ELMOD2 Fusion Protein Ag3653 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELMOD2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction