missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | ELOVL4 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896455
|
Proteintech
55023-1-AP-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886455
|
Proteintech
55023-1-AP-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to ELOVL4.
This gene encodes a membrane-bound protein which is a member of the ELO family, proteins which participate in the biosynthesis of fatty acids. Consistent with the expression of the encoded protein in photoreceptor cells of the retina, mutations and small deletions in this gene are associated with Stargardt-like macular dystrophy (STGD3) and autosomal domit Stargardt-like macular dystrophy (ADMD), also referred to as autosomal domit atrophic macular degeneration.Specifications
ELOVL4 | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q9EQC4, Q9GZR5 | |
315851, 6785, 83603 | |
Peptide | |
Primary | |
-20°C | |
ELOVL4 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ADMD, ELOVL4, STGD2, STGD3 | |
Elovl4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ELOVL4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech