Learn More
Merck MultiScreen™ Multiple Punch and Punch Tips
For rdionucleotide counting by transfer of individual filters to tubes
6720.00 SEK - 70700.00 SEK
Specifications
For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | ||||||
10156100
|
Merck
MADP19650 |
Punch tip |
6720.00 SEK
Pack of 50 |
Estimated Shipment: 07-08-2025
Log in to see stock. |
||||||
11546365
|
Merck
MAMP09608 |
Punch kit |
70700.00 SEK
Pack of 10 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
||||||
Description
- Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
- Remove filters from wells separately, eliminating cross-contamination
- Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
- Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifications
Radionucleotide counting by transfer of individual filters to tubes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Merck MultiScreen™ Multiple Punch and Punch Tips