Learn More
Merck Millipore Pure Water Storage Reservoir
Choose a Pure Water Storage Reservoir to store purified water without compromising quality.
Description
Description
Choose a Pure Water Storage Reservoir to store purified water without compromising quality.
Key Features
  • Polyethylene construction minimizes extractables that can contaminate water
  • Cylindrical shape minimizes area in contact with water
  • Smooth inner surface prevents biofilm formation
  • Conical bottom allows complete drainage for effective cleaning and sanitization
  • Hermetically sealed lid on large reservoir opening prevents air from leaking into reservoir and facilitates manual sanitization
Filling and Dispensing Features
  • Sensor-rod float triggers automatic reservoir refill
  • Water is fed into bottom of reservoir to prevent absorption of CO2
  • Two bottom valves allow direct connection to water polishing system and glassware washer
  • Water dispenses through valve on front of reservoir
Optional Features
  • Vent filter (available separately) guards against airborne impurities
  • 30, 60 and 100L reservoirs can be wall-mounted (require TANK FIX 01 wall mounting bracket)
  • Automatic Sanitization Module (A.S.M.) available separately to reduce bacteria levels
  • Optional distribution pump continuously delivers water under pressure
  • Water detector sensor (available separately) detects water on most surfaces such as floors and lab bench
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merck Millipore Pure Water Storage Reservoir