Learn More
EMG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11965-1-AP-150UL
 View more versions of this product
Product Code. 16889383
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
Specifications
Specifications
EMG1
Polyclonal
Unconjugated
EMG1
C2F, EMG1, Grcc2f, NEP1, Nucleolar protein EMG1 homolog, Protein C2f
Rabbit
Antigen Affinity Chromatography
RUO
10436, 14791, 312706
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35130, Q92979
EMG1
EMG1 Fusion Protein Ag2575
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EMG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated