missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EML2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13529-1-AP-20UL
This item is not returnable.
View return policy
Description
May modify the assembly dynamics of microtubules, such that microtubules are slightly longer, but more dynamic.Specifications
EML2 | |
Polyclonal | |
Unconjugated | |
EML2 | |
ELP70, EMAP 2, EMAP2, EMAPL2, EML2, HuEMAP 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24139, 72205 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95834, Q7TNG5 | |
Eml2 | |
EML2 Fusion Protein Ag4423 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
EML2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction