missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Emx1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-86500
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Emx1 Polyclonal specifically detects Emx1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifikationer
| Emx1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EMX1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RSLYGGPELVFPEAMNHPALTVHPAHQLGASPLQPPHSFFGAQHRDPLHFYPWVLRNRFFGHRFQASDVPQDGLLLHGPFARKPKRI | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| empty spiracles homeobox 1, empty spiracles-like protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2016 | |
| Human, Mouse | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering